tert-Butyl piperidine-4-carboxylate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A128268-100G
A128268-10G
A128268-25G
A128268-1G
A128268-5G
103801-100EA
5
USD
103801-100
76767-020
76767-022
77298-280
77313-404
77313-406
tert-Butyl piperidine-4-carboxylate hydrochloride 97%
tert-Butyl piperidine-4-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₂₀ClNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03788468 CAS Number: 892493-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...