tert-Butyl piperidin-4-ylcarbamate hydrochloride 95%

Supplier: Ambeed
Warning

A413223-1G A413223-5G A413223-250MG
103827-258EA 17 USD
103827-258 103838-304 76841-858 76841-860 76979-176
tert-Butyl piperidin-4-ylcarbamate hydrochloride 95%
tert-Butyl piperidin-4-ylcarbamate hydrochloride

Precaución: For research use only

Formula: C₁₀H₂₁ClN₂O₂
MW: 236.74 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03093509
CAS Number: 179110-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR