(±)-3-(Boc-amino)piperidine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl piperidin-3-ylcarbamate, 3-Aminopiperidine, 3-BOC protected

A111020-100G A111020-10G A111020-25G A111020-1G A111020-5G
103797-776EA 5 USD
103797-776 103823-582 76765-808 76765-810 76765-812 77310-390 77310-392
(±)-3-(Boc-amino)piperidine 98%
(±)-3-(Boc-amino)piperidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 304.8 °C (760 mm Hg)
Melting Pt: 102…108 °C
Flash Pt: 138.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839941
CAS Number: 172603-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR