O-tert-Butyl-N,N'-diisopropylisourea 95% (by titrimetric analysis)

Supplier: Ambeed
Warning

Synonyms: tert-Butyl N,N'-diisopropylcarbamimidate, 2-tert-Butyl-1,3-diisopropylisourea

A109935-100G A109935-10G A109935-250MG A109935-25G A109935-500G A109935-1G A109935-5G
103797-590EA 9 USD
103797-590 103823-396 77270-820 77270-822 77270-824 77270-826 77270-828 77310-202 77310-204
O-tert-Butyl-N,N'-diisopropylisourea 95% (by titrimetric analysis)
O-tert-Butyl-N,N'-diisopropylisourea

Precaución: for Research Use Only

Formula: C₁₁H₂₄N₂O
MW: 200.33 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06657672
CAS Number: 71432-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR