3-(Boc-amino)propylbromide 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(tert-Butoxycarbonylamino)propyl bromide, 3-(Boc-amino)propyl bromide, tert-Butyl N-(3-Bromopropyl)carbamate, N-(3-Bromopropyl)carbamic acid tert-Butyl ester
A221662-10G
A221662-1G
A221662-100G
A221662-500G
103818-940EA
5
USD
103818-940
103849-818
77165-234
77303-608
77550-434
77683-639
3-(Boc-amino)propylbromide 98%, Ambeed.Inc
3-(Boc-amino)propylbromide
Precaución: For research use only.
Formula:
C8H16BrNO2 MW: 238.1221 g/mol Melting Pt: 37…39 °C Storage Temperature: Freezer |
MDL Number:
MFCD02683429 CAS Number: 83948-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...