tert-Butyl methylsulfonylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl methylsulphonylcarbamate

A215124-10G A215124-250MG A215124-25G A215124-5G
103818-456EA 24 USD
103818-456 103849-160 76826-014 76826-016 77087-686 77087-688
tert-Butyl methylsulfonylcarbamate 97%
tert-Butyl methylsulfonylcarbamate

Precaución: for Research Use Only

Formula: C₆H₁₃NO₄S
MW: 195.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13194912
CAS Number: 147751-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR