tert-Butyl-N-methylcarbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl methylcarbamate
A127959-100G
A127959-10G
A127959-500G
A127959-1G
A127959-25G
A127959-5G
103801-042EA
5
USD
103801-042
76811-022
76811-024
76811-026
77298-256
77313-318
77313-320
tert-Butyl-N-methylcarbamate 97%
tert-Butyl-N-methylcarbamate
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ Boiling Pt: 176.6 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD08899404 CAS Number: 16066-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...