tert-Butyl methyl(piperidin-4-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl methyll(4-piperidyl)carbamate, Methyl(4-piperidyl)carbamic acid tert-Butyl ester
A191686-100G
A191686-250MG
A191686-1G
A191686-25G
A191686-5G
A191686-10G
103816-392EA
11
USD
103816-392
103846-996
76800-000
76800-002
77084-850
77084-852
77084-854
77593-222
tert-Butyl methyl(piperidin-4-yl)carbamate 97%
tert-Butyl methyl(piperidin-4-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol Boiling Pt: 287.9 °C (760 mm Hg) Flash Pt: 127.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02683051 CAS Number: 108612-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...