tert-Butyl methyl(4-oxocyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

A130557-1G A130557-25G A130557-5G
103801-440EA 12 USD
103801-440 77313-882 77313-884 77313-886
tert-Butyl methyl(4-oxocyclohexyl)carbamate 97%
tert-Butyl methyl(4-oxocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3000 g/mol
Boiling Pt: 316.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09037870
CAS Number: 400899-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR