tert-Butyl methyl malonate 97%

Supplier: Ambeed
Warning

Synonyms: tert.-Butyl methyl malonate

A396101-100G A396101-10G A396101-500G A396101-1G A396101-25G A396101-5G
103826-978EA 9 USD
103826-978 103838-016 76788-896 76788-898 76788-900 76841-438 76841-440 76841-442
tert-Butyl methyl malonate 97%
tert-Butyl methyl malonate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.19 g/mol
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00042856
CAS Number: 42726-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR