tert-Butyl isonicotinate 97%

Supplier: Ambeed
Warning

A165039-100MG A165039-250MG A165039-25G A165039-1G A165039-5G
103805-936EA 22 USD
103805-936 103844-194 76647-198 76647-200 76647-202 77300-756 77300-758
tert-Butyl isonicotinate 97%
tert-Butyl isonicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19600069
CAS Number: 81660-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR