tert-Butyl carbazate 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl hydrazinecarboxylate

A245155-25G A245155-10G
103809-744EA 232 USD
103809-744 77090-694 77288-838
tert-Butyl carbazate 98%
tert-Butyl carbazate

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O₂
MW: 132.16 g/mol
Boiling Pt: 303.6 °C (760 mm Hg)
Melting Pt: 39…42 °C
Flash Pt: 92 °C
Storage Temperature: Freezer
MDL Number: MFCD00007593
CAS Number: 870-46-2
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR