tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Octahydropyrrolo[3,4-c]pyrrole, N2-BOC protected

A189902-250MG A189902-1G A189902-25G A189902-5G
103816-232EA 29 USD
103816-232 103846-828 77012-256 77084-600 77084-602 77084-604
tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate 97%
tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.2887 g/mol
Boiling Pt: 293 °C (760 mm Hg)
Melting Pt: 35…38 °C
Flash Pt: 132.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115128
CAS Number: 141449-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR