tert-Butyl hex-5-yn-1-ylcarbamate 95%

Supplier: Ambeed
Warning

A141036-100MG A141036-10G A141036-250MG A141036-5G A141036-25G
103802-938EA 87 USD
103802-938 77095-438 77095-440 77095-442 77299-186 77685-206
tert-Butyl hex-5-yn-1-ylcarbamate 95%
tert-Butyl hex-5-yn-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₂
MW: 197.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18839148
CAS Number: 151978-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR