tert-Butyl ethyl malonate 97%
Supplier: Ambeed
|
Warning
|
A172679-25G
A172679-100G
A172679-10G
A172679-500G
A172679-5G
103845-030EA
6
USD
103845-030
77082-236
77259-956
77259-958
77259-960
77301-212
tert-Butyl ethyl malonate 97%
tert-Butyl ethyl malonate
Precaución: for Research Use Only
Formula:
C₉H₁₆O₄ Boiling Pt: 210.7 °C (760 mm Hg) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009193 CAS Number: 32864-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...