N-Boc-endo-3-aminotropane 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl endo-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate, tert-Butyl (1S,5R)-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate

A114194-100MG A114194-10G A114194-250MG A114194-25G A114194-1G A114194-5G
103798-366EA 28 USD
103798-366 103824-206 76971-078 76971-080 76971-082 76971-084 77311-024 77311-026
N-Boc-endo-3-aminotropane 96%
N-Boc-endo-3-aminotropane

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.315 g/mol
Boiling Pt: 313.2 °C (760 mm Hg)
Flash Pt: 143.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18428077
CAS Number: 207405-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR