(2R,6S)-tert-Butyl-2,6-dimethylpiperazine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl cis-2,6-dimethylpiperazine-1-carboxylate
A112727-250MG
A112727-1G
A112727-5G
103798-090EA
21
USD
103798-090
103823-928
76821-944
77310-748
77310-750
(2R,6S)-tert-Butyl-2,6-dimethylpiperazine-1-carboxylate 97%
(2R,6S)-tert-Butyl-2,6-dimethylpiperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09265026 CAS Number: 180975-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...