tert-Butyl carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Carbamic acid tert-butyl ester

A200170-100G A200170-10G A200170-500G A200170-1000G A200170-25G
103809-340EA 147 USD
103809-340 76785-296 76785-298 76785-300 76785-302 77085-858 77085-860
tert-Butyl carbamate 98%
tert-Butyl carbamate

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 195.9 °C (760 mm Hg)
Melting Pt: 105…108 °C
Flash Pt: 82.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007962
CAS Number: 4248-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR