tert-Butyl but-3-yn-1-ylcarbamate 95%

Supplier: Ambeed
Warning

A177840-10G A177840-250MG A177840-1G A177840-25G A177840-5G
103807-272EA 23 USD
103807-272 103845-606 77029-316 77029-318 77082-998 77083-000 77083-002
tert-Butyl but-3-yn-1-ylcarbamate 95%
tert-Butyl but-3-yn-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₂
MW: 169.221 g/mol
Flash Pt: 105.326 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698150
CAS Number: 149990-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR