tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl bis(2-hydroxyethyl)carbamate, N-BOC-Diethanolamine

A134492-100G A134492-10G A134492-25G A134492-500G A134492-1G A134492-5G
103802-016EA 9 USD
103802-016 76811-410 76811-412 76811-414 76811-416 77298-714 77314-776
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate 97%
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₉NO₄
MW: 205.26 g/mol
Boiling Pt: 324.2 °C (760 mm Hg)
Flash Pt: 149.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862953
CAS Number: 103898-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR