tert-Butyl bis(2-chloroethyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: N-BOC-N,N-Bis(2-chloroethyl)amine

A295462-10G A295462-25G A295462-5G A295462-100G
103814-058EA 5 USD
103814-058 103835-192 76723-556 76837-848 76837-850 77879-583
tert-Butyl bis(2-chloroethyl)carbamate 97%
tert-Butyl bis(2-chloroethyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₇Cl₂NO₂
Boiling Pt: 290.6 °C (760 mm Hg)
Flash Pt: 129.6 °C
Storage Temperature: Freezer
MDL Number: MFCD06858679
CAS Number: 118753-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR