tert-Butyl aziridine-1-carboxylate 95%

Supplier: Ambeed
Danger

A330357-1G A330357-5G A330357-250MG
103825-550EA 242 USD
103825-550 103836-510 76839-486 76839-488 77571-950
tert-Butyl aziridine-1-carboxylate 95%
tert-Butyl aziridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.18 g/mol
Boiling Pt: 174.8±9.0 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD16036961
CAS Number: 97308-23-1
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR