tert-Butyl azetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A226506-250MG A226506-1G A226506-5G A226506-100MG
103819-252EA 35 USD
103819-252 103850-156 76815-052 77088-886 77088-888 77671-994
tert-Butyl azetidine-1-carboxylate 97%
tert-Butyl azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 198.7±9.0 °C (760 mm Hg)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264397
CAS Number: 147621-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR