tert-Butyl azetidin-3-ylcarbamate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Boc-amino)azetidine hydrochloride

A129531-100G A129531-10G A129531-25G A129531-1G A129531-5G
103801-298EA 7 USD
103801-298 76782-252 76782-254 76782-256 77313-680 77313-682
tert-Butyl azetidin-3-ylcarbamate hydrochloride 97%
tert-Butyl azetidin-3-ylcarbamate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₇ClN₂O₂
MW: 208.69 g/mol
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09839423
CAS Number: 217806-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR