tert-Butyl azetidin-3-ylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Boc-amino)azetidine

A177395-10G A177395-250MG A177395-25G A177395-1G A177395-5G
103807-232EA 72 USD
103807-232 103845-560 76872-624 76872-626 77082-952 77301-506 77301-508
tert-Butyl azetidin-3-ylcarbamate 97%
tert-Butyl azetidin-3-ylcarbamate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
Boiling Pt: 267.5 °C (760 mm Hg)
Melting Pt: 63…72 °C
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01861759
CAS Number: 91188-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR