tert-Butyl-N-allylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate

A100103-100G A100103-10G A100103-1G A100103-25G A100103-500G A100103-5G
103820-972EA 18 USD
103820-972 76780-340 76780-342 76780-344 76780-346 76780-348 77296-570
tert-Butyl-N-allylcarbamate 98%
tert-Butyl-N-allylcarbamate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Melting Pt: 36…38 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00191870
CAS Number: 78888-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR