3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 9-oxo-3-azaspiro[5.5]undecane-3-carboxylate

A180994-100MG A180994-250MG A180994-1G A180994-5G A180994-10G
103807-580EA 43 USD
103807-580 103845-942 76985-758 76985-760 77301-704 77301-706 77611-754
3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%
3-Boc-9-oxo-3-azaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C₁₅H₂₅NO₃
MW: 267.37 g/mol
Boiling Pt: 381.1 °C (760 mm Hg)
Melting Pt: 70…73 °C
Flash Pt: 184.3 °C
Storage Temperature: Ambient
MDL Number: MFCD12198558
CAS Number: 873924-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR