Amino-PEG2-acid tert-butyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 3-[2-(2-Aminoethoxy)ethoxy]propanoate, 3-[2-(2-Aminoethoxy)ethoxy]propanoic acid tert-butyl ester, 9-Amino-4,7-dioxanonanoic acid tert-butyl ester, NH2-PEG2-C2-Boc, tert-Butyl 9-Amino-4,7-dioxanonanoate, Amino-dPEG®2-t-butyl ester
A312281-1G
A312281-5G
A312281-100MG
A312281-250MG
103814-696EA
15
USD
103814-696
103835-886
76838-690
76838-692
76875-778
76875-780
Amino-PEG2-acid tert-butyl ester 95%
Amino-PEG2-acid tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₁H₂₃NO₄ MW: 233.305 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11041135 CAS Number: 756525-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...