7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

A180310-100MG A180310-250MG A180310-1G A180310-5G
103807-530EA 38 USD
103807-530 103845-884 76824-856 76824-858 77301-668 77301-670
7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester 95%
7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD18325190
CAS Number: 1234616-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR