tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
A110934-100G
A110934-10G
A110934-250MG
A110934-25G
A110934-1G
A110934-5G
103797-758EA
6
USD
103797-758
103823-566
76970-766
76970-768
76970-770
76970-772
77310-372
77310-374
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate 97%
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol Boiling Pt: 272.5±23.0 °C (760 mm Hg) Flash Pt: 118.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD12407050 CAS Number: 161157-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...