tert-Butyl-7-amino-3,4-dihydroquinoline-1(2H)-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 7-amino-3,4-dihydroquinoline-1(2H)-carboxylate

A333439-1G A333439-5G
103825-646EA 131 USD
103825-646 103836-612 77306-454 77306-456
tert-Butyl-7-amino-3,4-dihydroquinoline-1(2H)-carboxylate 97%
tert-Butyl-7-amino-3,4-dihydroquinoline-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.3200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566742
CAS Number: 351324-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR