tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-oxo-2-azaspiro[3.3]heptane-2-carboxylate

A175251-100MG A175251-10G A175251-250MG A175251-1G A175251-25G A175251-5G A175251-100G A175251-500G
103807-016EA 17 USD
103807-016 103845-320 76784-392 76784-394 76784-396 77082-658 77082-660 77082-662 77593-104 77667-434
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate 98%
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MW: 211.2576 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15071430
CAS Number: 1181816-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR