tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylic acid tert-Butyl ester, tert-Butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate, 1-(tert-Butoxycarbonyl)-3,4-epoxypyrrolidine, 1-Boc-3,4-epoxypyrrolidine
A145116-100G
A145116-10G
A145116-25G
A145116-1G
A145116-5G
103803-496EA
9
USD
103803-496
103841-576
77272-710
77272-712
77272-714
77316-846
77316-848
tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate 97%
tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₅NO₃ Boiling Pt: 252.7 °C (760 mm Hg) Flash Pt: 106.644 °C Storage Temperature: Ambient |
MDL Number:
MFCD08691407 CAS Number: 114214-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...