6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxy-2-azaspiro[3.3]heptane, N-BOC protected, tert-Butyl 6-hydroxy-2-azaspiro[3.3]heptane-2-carboxylate

A331968-100MG A331968-10G A331968-250MG A331968-25G A331968-5G A331968-100G
103825-594EA 14 USD
103825-594 103836-562 76788-176 76788-178 76788-180 76839-556 76839-558 77668-502
6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester 95%
6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
Melting Pt: 127…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15071431
CAS Number: 1147557-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR