tert-Butyl 6-chloronicotinate 95%

Supplier: Ambeed
Warning

A405604-1G A405604-5G A405604-250MG
103827-122EA 27 USD
103827-122 103838-162 76841-650 76841-652 77033-708
tert-Butyl 6-chloronicotinate 95%
tert-Butyl 6-chloronicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
Boiling Pt: 270.7 °C (760 mm Hg)
Melting Pt: 53…54 °C
Flash Pt: 117.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08277247
CAS Number: 115309-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR