tert-Butyl 6-bromoindoline-1-carboxylate 95%

Supplier: Ambeed
Warning

A109557-100MG A109557-250MG A109557-1G
103797-528EA 121 USD
103797-528 103823-328 77008-854 77008-856 77310-140
tert-Butyl 6-bromoindoline-1-carboxylate 95%
tert-Butyl 6-bromoindoline-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₂
MW: 298.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059279
CAS Number: 214614-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR