tert-Butyl-6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate, 6-Amino-2-N-BOC-1,2,3,4-tetrahydro-isoquinoline

A116941-100MG A116941-10G A116941-250MG A116941-25G A116941-1G A116941-5G
103798-804EA 36 USD
103798-804 103824-676 76781-436 76781-438 76781-440 76781-442 77297-502 77311-464
tert-Butyl-6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate 98%
tert-Butyl-6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.3200 g/mol
Boiling Pt: 394.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03426351
CAS Number: 164148-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR