tert-Butyl-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

A806688-250MG A806688-1G
103841-072EA 86 USD
103841-072 76791-650 76845-320
tert-Butyl-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate 98%
tert-Butyl-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂S
MW: 239.3300 g/mol
Boiling Pt: 339.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11518948
CAS Number: 230301-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR