tert-Butyl 5-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate 97%

Supplier: Ambeed
Warning

A220932-250MG A220932-5G
103818-880EA 74 USD
103818-880 103849-760 76814-882 77303-566
tert-Butyl 5-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate 97%
tert-Butyl 5-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MW: 211.26 g/mol
Boiling Pt: 308 °C (760 mm Hg)
Flash Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09040745
CAS Number: 198835-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR