tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Homopiperazin-5-one, N1-BOC protected, tert-Butyl 5-oxo-1,4-diazepane-1-carboxylate

A252205-10G A252205-250MG A252205-5G
103812-032EA 24 USD
103812-032 103851-874 77000-708 77000-710 77091-284
tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate 97%
tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.2615 g/mol
Boiling Pt: 373.1 °C (760 mm Hg)
Melting Pt: 168…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115319
CAS Number: 190900-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR