tert-Butyl 5-methyl-2,4-dioxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A516842-100MG A516842-10G A516842-250MG A516842-1G A516842-5G
103828-464EA 417 USD
103828-464 103839-570 77169-470 77169-472 77169-474 77307-682 77307-684
tert-Butyl 5-methyl-2,4-dioxopiperidine-1-carboxylate 95%
tert-Butyl 5-methyl-2,4-dioxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.25 g/mol
Boiling Pt: 376.3±35.0 °C (760 mm Hg)
Flash Pt: 181.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD24466520
CAS Number: 942425-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR