tert-Butyl 5-hydroxyhexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A168950-5G
A168950-250MG
103806-332EA
73
USD
103806-332
103844-588
77081-658
77163-432
tert-Butyl 5-hydroxyhexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate 95%
tert-Butyl 5-hydroxyhexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₃ MW: 227.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD15071646 CAS Number: 203663-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...