tert-Butyl-5-bromopicolinate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-bromopicolinate, 5-Bromo-pyridine-2-carboxylic acid t-butyl ester, tert-Butyl 5-bromopyridine-2-carboxylate

A228900-250MG A228900-1G A228900-5G
103819-426EA 29 USD
103819-426 103850-332 76815-112 77089-142 77089-144
tert-Butyl-5-bromopicolinate 97%
tert-Butyl 5-bromopicolinate

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
MW: 258.1100 g/mol
Boiling Pt: 317.6 °C (760 mm Hg)
Melting Pt: 92…94 °C
Flash Pt: 145.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08436052
CAS Number: 845306-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR