tert-Butyl-5-bromoisoindoline-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-bromoisoindoline-2-carboxylate

A146414-10G A146414-250MG A146414-1G A146414-5G
103803-664EA 19 USD
103803-664 103841-754 76783-170 76783-172 77317-078 77317-080
tert-Butyl-5-bromoisoindoline-2-carboxylate 95%
tert-Butyl-5-bromoisoindoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₂
MW: 298.1800 g/mol
Boiling Pt: 354.7 °C (760 mm Hg)
Flash Pt: 168.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09953049
CAS Number: 201940-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR