tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

A179819-100MG A179819-10G A179819-250MG A179819-1G
103807-474EA 106 USD
103807-474 103845-826 76784-542 76784-544 76784-546 77083-300
tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁BrN₂O₃
MW: 381.2600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566052
CAS Number: 873779-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR