tert-Butyl 5-bromo-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A142562-100MG A142562-250MG
103803-182EA 306 USD
103803-182 77258-896 77258-898
tert-Butyl 5-bromo-1H-pyrazole-3-carboxylate 97%
tert-Butyl 5-bromo-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O₂
MW: 247.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30067292
CAS Number: 2089292-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR