1-Boc-5-bromoindole 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-5-bromoindole, 5-Bromo-1-tert-butoxycarbonylindole, tert-Butyl 5-bromo-1H-indole-1-carboxylate

A138774-10G A138774-250MG A138774-1G A138774-25G A138774-5G
103802-664EA 10 USD
103802-664 76782-822 76782-824 77315-722 77315-724 77315-726
1-Boc-5-bromoindole 97%
1-Boc-5-bromoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
Boiling Pt: 367.9 °C (760 mm Hg)
Melting Pt: 56…57 °C
Storage Temperature: Ambient
MDL Number: MFCD02090067
CAS Number: 182344-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR