3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5,6-dihydropyridine-1(2H)-carboxylate, 1-Boc-1,2,3,6-tetrahydropyridine, tert-Butyl 1,2,3,6-Tetrahydropyridine-1-carboxylate, 1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1,2,3,6-Tetrahydropyridine-1-carboxylic acid tert-Butyl ester

A303732-25G A303732-5G A303732-100G A303732-1G
103814-390EA 5 USD
103814-390 103835-556 76838-236 76838-238 77114-516 77305-884
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
Boiling Pt: 241.5 °C (760 mm Hg)
Flash Pt: 99.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04972245
CAS Number: 85838-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR