tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindoline-2-carboxylate 97%

Supplier: Ambeed
Warning

A267611-100MG A267611-250MG A267611-5G
103812-868EA 138 USD
103812-868 103852-768 76787-126 76787-128 77304-984
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindoline-2-carboxylate 97%
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₈BNO₄
MW: 345.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226842
CAS Number: 905273-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR