tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole-1-carboxylate 97%

Supplier: Ambeed
Warning

A334262-100MG A334262-250MG A334262-1G
103825-654EA 428 USD
103825-654 76989-312 76989-314 77306-464
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole-1-carboxylate 97%
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₅BN₂O₄
MW: 344.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24038762
CAS Number: 1021918-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR